Learn More
Research Products International Corp Manganese Chloride 1M Solution, 50 Milliliters
SDP
Catalog No. 50997913
Encompass_Preferred
Manganese Chloride is used for dC and dG tailing of DNA.
Catalog No. 50-997-913 Supplier Research Products International Corp Supplier No. M2010050.0
Manganese Chloride is used for dC and dG tailing of DNA.