Learn More
Alkali Scientific Manganese Chloride 1M Solution, 50 Milliliters
SDP
Supplier:  Alkali Scientific ANG002CP
Encompass_Preferred
Manganese Chloride 1M Solution is a laboratory reagent used for various chemical and biochemical applications, particularly as a catalyst or in enzyme activation studies. The 50-milliliter volume of this solution provides a convenient format for adding a precise amount of manganese to experiments requiring high concentrations of this essential metal ion. Manganese plays a key role in biological processes such as oxidation-reduction reactions and enzyme catalysis. This solution is ideal for use in research involving enzyme assays, redox reactions, and other biochemical processes that require precise manganese levels for proper activation or reaction kinetics.
Catalog No. 50-006-60195