Learn More
Medchemexpress LLC Manganese chloride | 7773-01-5 | MFCD00011114 | 99.1% | 125.84 g/mol | Cl2Mn | 500 G
SDP
Supplier:  Medchemexpress LLC HYW019894500G
Encompass_Preferred
Manganese chloride (Cl2Mn, CAS 7773-01-5) is an inorganic manganese(II) salt supplied for research and biochemical applications. Product literature describes it as an orally active MRI liver contrast agent and reports biological activity including modulation of mTOR signaling and induction of apoptosis; it has been used in studies of cognitive impairment, hematopoietic recovery, radiation-induced tissue damage, and renal impairment. Molecular weight 125.84 g/mol.
  • Inorganic manganese(II) salt, water-soluble reagent.
  • Used as an orally active MRI liver contrast agent in research.
  • Modulates mTOR signaling and can induce apoptosis.
  • Applied in studies of cognitive impairment and hematopoietic recovery.
  • Available in larger pack sizes suitable for batch experiments.
Catalog No. 50-003-00994