Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Manganese Chloride ACS AR, Macron Fine Chemicals™

Catalog No. MK6126500 Shop All Avantor Products
Change view
Click to view available options
Quantity:
2.5 kg
4 x 500 g
12 x 125 g
Packaging:
Glass
Poly
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
MK-6126-500 4 x 500 g Glass
MK-6126-125 12 x 125 g Glass
MK-6126-212 2.5 kg Poly
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. MK-6126-500 Supplier Avantor Supplier No. 612612

missing translation for 'validMainframeMsg'

Chemical Identifiers

CAS 13446-34-9
Molecular Formula Cl2H8MnO4
Molecular Weight (g/mol) 197.90
MDL Number MFCD00149792
InChI Key CNFDGXZLMLFIJV-UHFFFAOYSA-L
IUPAC Name manganese(2+) tetrahydrate dichloride
SMILES O.O.O.O.[Cl-].[Cl-].[Mn++]

Specifications

Quantity 4 x 500 g
Formula Weight 197.91
Grade ACS
Packaging Glass
Chemical Name or Material Manganese Chloride, 4-Hydrate
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.