Learn More
Medchemexpress LLC Manganese chloride (tetrahydrate), molecular biology grade | 13446-34-9 | 99.1% | 197.91 | 10 MG
SDP
Catalog No. 5000344897
Encompass_Preferred
Manganese chloride (tetrahydrate), molecular biology grade, ≥99.0% (KT) is an orally active and blood-brain barrier penetrant compound. It influences the activities of multiple enzymes in cells, including histone acetyltransferase (HAT) and histone deacetylase (HDAC), thereby affecting gene expression. It exhibits neurotoxicity, embryotoxicity, and reproductive toxicity, and is primarily used in neurodegenerative diseases and toxicology research.
  • Affects activities of multiple enzymes in cells, such as regulating HAT and HDAC, influencing gene expression.
  • Exhibits neurotoxicity, embryotoxicity, and reproductive toxicity.
  • Mainly used in neurodegenerative diseases and toxicology research.
  • For research use only.
Catalog No. 50-003-44897 Supplier Medchemexpress LLC Supplier No. HY109521A5G
Manganese chloride (tetrahydrate), molecular biology grade, ≥99.0% (KT) is an orally active and blood-brain barrier penetrant compound. It influences the activities of multiple enzymes in cells, including histone acetyltransferase (HAT) and histone deacetylase (HDAC), thereby affecting gene expression. It exhibits neurotoxicity, embryotoxicity, and reproductive toxicity, and is primarily used in neurodegenerative diseases and toxicology research.
  • Affects activities of multiple enzymes in cells, such as regulating HAT and HDAC, influencing gene expression.
  • Exhibits neurotoxicity, embryotoxicity, and reproductive toxicity.
  • Mainly used in neurodegenerative diseases and toxicology research.
  • For research use only.