Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Manganese Dioxide (Manganese (IV) Oxide), Reagents

Catalog No. 01337182
Change view
Click to view available options
Quantity:
100 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
01-337-182 100 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 01-337-182 Supplier Reagents Holdings Llc Supplier No. C2242500100C2
 Restricted Item
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Manganese Dioxide (Manganese (IV) Oxide), Chemically Pure, for general laboratory use. Manufactured in ISO 9001 facility.

Chemical Identifiers

CAS 1313-13-9
Molecular Formula MnO2
Molecular Weight (g/mol) 86.94
MDL Number MFCD00003463
InChI Key NUJOXMJBOLGQSY-UHFFFAOYSA-N
Synonym Manganese oxide, Manganese monoxide, Manganosite
IUPAC Name dioxomanganese
SMILES O=[Mn]=O

Specifications

Melting Point 535°C
Color Black
Physical Form Solid
Quantity 100 g
Grade Pure
Packaging Glass Bottle
Appearance Black
Specific Gravity 5.08
Chemical Name or Material Manganese Dioxide (Manganese (IV) Oxide)
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.