Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

LaMotte™ Manganese HR SMART 3 Reagent System
SDPSCI_ED

Catalog No. S95013 Shop All LaMotte Company Products
Change view
Click to view available options
No. of Tests:
50
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. No. of Tests
S95013 50
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. S95013 Supplier LaMotte™ Supplier No. 3669SC
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Manganese high range reagent system for use with SMART 3 Colorimeter.

  • Range 0.3 to 15.0 ppm
  • 50 Tests
  • Periodate Method

Specifications

pH Range 3 to 10
For Use With (Application) For Industrial Waters
For Use With (Equipment) SMART 3 Colorimeter
No. of Tests 50
Range 0.3 to 15.0 ppm
Test Method Periodate Method
Type Reagent System
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.