Learn More
Strem, An Ascensus Company CAS# 6156-78-1. 5g. Manganese(II) acetate tetrahydrate (99.999%-Mn) PURATREM. MFCD00062552
SDP
Catalog No. 5090110903
Encompass
CAS# 6156-78-1. 5g. Manganese(II) acetate tetrahydrate (99.999%-Mn) PURATREM. MFCD00062552. Molecular Weight: 173.02 (245.08). Molecular Formula: Mn(CH3COO)2∙4H2O. Color/form: pale pink xtl. Strem# 25-0760. http://www.strem.com/catalog/v/25-0760/
Catalog No. 50-901-10903 Supplier Strem, An Ascensus Company Supplier No. 2507605G
CAS# 6156-78-1. 5g. Manganese(II) acetate tetrahydrate (99.999%-Mn) PURATREM. MFCD00062552. Molecular Weight: 173.02 (245.08). Molecular Formula: Mn(CH3COO)2∙4H2O. Color/form: pale pink xtl. Strem# 25-0760. http://www.strem.com/catalog/v/25-0760/