Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Manganese(II) bromide, anhydrous, 99%, H{2}O <0.3%

Catalog No. AA3273609
Change view
Click to view available options
Quantity:
10 g
50 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AA3273609 10 g
AA3273618 50 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AA3273609 Supplier Thermo Scientific Chemicals Supplier No. 03273609
Add to Cart

CAS: 13446-03-2

It can be used in place of palladium in the Stille reaction, which couples two carbon atoms using an organotin compound.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 13446-03-2
MDL Number MFCD00011113

Specifications

Melting Point 698°C
Physical Form Powder
Quantity 10 g
Merck Index 14,5725
Solubility Information Soluble in water. (1273g/L)
Formula Weight 214.76
Percent Purity 99%
Concentration or Composition (by Analyte or Components) H2O <0.3%
Odor Odorless
Sensitivity Hygroscopic
Density 4.38 g/mL
Chemical Name or Material Manganese(II) bromide, Anhydrous
Show More Show Less
Hazard Category H302+H312+H332
Hazard Statement GHS H Statement
H302
Harmful if swallowed.
Precautionary Statement P261-P264b-P270-P271-P280-P301+P312-P302+P352-P304+P340-P312-P330-P363-P501c
EINECSNumber 236-591-9
TSCA Yes
Recommended Storage Ambient temperatures

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.