Learn More
Strem, An Ascensus Company CAS# 598-62-9. 500g. Manganese(II) carbonate, min. 90%. MFCD00011116
SDP
Catalog No. 5090114617
Encompass
CAS# 598-62-9. 500g. Manganese(II) carbonate, min. 90%. MFCD00011116. Molecular Weight: 114.95. Molecular Formula: MnCO3. Color/form: light brown pwdr. Strem# 93-2533. http://www.strem.com/catalog/v/93-2533/
Catalog No. 50-901-14617 Supplier Strem, An Ascensus Company Supplier No. 932533500G
CAS# 598-62-9. 500g. Manganese(II) carbonate, min. 90%. MFCD00011116. Molecular Weight: 114.95. Molecular Formula: MnCO3. Color/form: light brown pwdr. Strem# 93-2533. http://www.strem.com/catalog/v/93-2533/