Learn More
Medchemexpress LLC Manganese(II) chloride | 7773-01-5 | MFCD00011114 | 99.5% | 125.84 g/mol | MnCl2 | 100 G
SDP
Catalog No. 5000299973
Encompass_Preferred
Manganese(II) chloride (MnCl2, CAS 7773-01-5) is a research-grade inorganic salt used as a biochemical reagent and in preclinical studies. It appears in literature as an MRI liver contrast agent and is used to study cellular signaling, apoptosis, hematopoietic recovery, and models of renal impairment. Typical assay values are reported on the manufacturer COA.
  • Used as a biochemical reagent in cellular and biochemical assays.
  • Investigated as an MRI liver contrast agent in preclinical research.
  • Influences apoptosis and mTOR signaling pathways.
  • Supports studies of hematopoietic recovery and radiation damage mitigation.
  • Storage: 4°C, sealed, away from moisture; in solution -80°C up to 6 months or -20°C up to 1 month.
  • High assay typically reported (~99.5% per COA).
Catalog No. 50-002-99973 Supplier Medchemexpress LLC Supplier No. HYW019894100G
Manganese(II) chloride (MnCl2, CAS 7773-01-5) is a research-grade inorganic salt used as a biochemical reagent and in preclinical studies. It appears in literature as an MRI liver contrast agent and is used to study cellular signaling, apoptosis, hematopoietic recovery, and models of renal impairment. Typical assay values are reported on the manufacturer COA.
  • Used as a biochemical reagent in cellular and biochemical assays.
  • Investigated as an MRI liver contrast agent in preclinical research.
  • Influences apoptosis and mTOR signaling pathways.
  • Supports studies of hematopoietic recovery and radiation damage mitigation.
  • Storage: 4°C, sealed, away from moisture; in solution -80°C up to 6 months or -20°C up to 1 month.
  • High assay typically reported (~99.5% per COA).