Learn More
Strem, An Ascensus Company CAS# 7773-01-5. 500g. Manganese(II) chloride, anhydrous, 97%. MFCD00011114
SDP
Catalog No. 5090114612
Encompass
CAS# 7773-01-5. 500g. Manganese(II) chloride, anhydrous, 97%. MFCD00011114. Molecular Weight: 125.84. Molecular Formula: MnCl2. Color/form: pink flakes Strem# 93-2527. http://www.strem.com/catalog/v/93-2527/
Catalog No. 50-901-14612 Supplier Strem, An Ascensus Company Supplier No. 932527500G
CAS# 7773-01-5. 500g. Manganese(II) chloride, anhydrous, 97%. MFCD00011114. Molecular Weight: 125.84. Molecular Formula: MnCl2. Color/form: pink flakes Strem# 93-2527. http://www.strem.com/catalog/v/93-2527/