Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Sigma Aldrich Manganese(II) Chloride Monohydrate

Catalog No. 1322025KGR Shop All MilliporeSigma Sigma Organic Chemistry Products
Encompass
Encompass_Preferred
Change view
Click to view available options
Quantity:
2.5 kg
6 x 2.5 kg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
13-220-25KGR 2.5 kg
50-308-1425 6 x 2.5 kg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Danger
Catalog No. 13-220-25KGR Supplier Sigma Aldrich Supplier No. 132202.5KGR
Add to Cart
Add to Cart
Danger

MilliporeSigma Sigma Organics products encompass a wide range of quality reagents, solvents, catalysts, and building blocks for organic synthesis. From benchtop discovery to process development and scale-up, Sigma Organics solutions are built to meet the needs of synthetic chemists.

Starting material for preparation of organomanganese chloride reagents; in combination with RLi or RMgX catalyzes reduction of vinyl and aryl halides, stereoselective coupling of vinyl iodides, and alkylation of enol triflates; also used for Mn-Cu-catalyzed conjugate addition of RMgX to enones and for Mn-catalyzed acylation of RMgX by acyl chlorides; in combination with NaBH4 is diastereoselective reducing agent; starting material for the preparation of Mn enolates regio- andstereo-selectively.

Specifications

CAS 64333-01-3
Molecular Formula Cl2Mn -+ H2O
Linear Formula MnCl2 -+ H2O
Quantity 2.5 kg
MDL Number MFCD00149793
Synonym Manganese dichloride monohydrate
Molecular Weight (g/mol) 143.86 g/mol
Biological Activity Brain
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.