Learn More
Strem, An Ascensus Company CAS# 13446-34-9. 250g. Manganese(II) chloride tetrahydrate, 98+% (ACS). MFCD00149792
SDP
Catalog No. 5090114618
Encompass
CAS# 13446-34-9. 250g. Manganese(II) chloride tetrahydrate, 98+% (ACS). MFCD00149792. Molecular Weight: 125.84 (197.91). Molecular Formula: MnCl2∙4H2O. Color/form: rose xtl. Strem# 93-2534. http://www.strem.com/catalog/v/93-2534/
Catalog No. 50-901-14618 Supplier Strem, An Ascensus Company Supplier No. 932534250G
CAS# 13446-34-9. 250g. Manganese(II) chloride tetrahydrate, 98+% (ACS). MFCD00149792. Molecular Weight: 125.84 (197.91). Molecular Formula: MnCl2∙4H2O. Color/form: rose xtl. Strem# 93-2534. http://www.strem.com/catalog/v/93-2534/