Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Manganese(II) chloride tetrahydrate, 99+%, for analysis

Catalog No. p-4509875
Change view
Click to view available options
Quantity:
100 g
500 g
2.5 kg
Packaging:
Plastic bottle
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
AC205891000 100 g Plastic bottle
AC205895000 500 g Plastic bottle
AC205890025 2.5 kg Plastic bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. AC205891000 Supplier Thermo Scientific Chemicals Supplier No. 205891000
Only null left
Add to Cart

CAS: 13446-34-9 | Cl2H8MnO4 | 197.90 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 13446-34-9
Molecular Formula Cl2H8MnO4
Molecular Weight (g/mol) 197.90
MDL Number MFCD00149792
InChI Key CNFDGXZLMLFIJV-UHFFFAOYSA-L
IUPAC Name manganese(2+) tetrahydrate dichloride
SMILES O.O.O.O.[Cl-].[Cl-].[Mn++]

Specifications

Melting Point 58.0°C
Color Pink
Physical Form Fine Crystalline Powder
Quantity 100 g
Assay Percent Range 99% min. (Complexometry)
Linear Formula MnCl2·4H2O
Fieser 07,222; 09,288
Merck Index 15,5793
Solubility Information Solubility in water: 1980g/L (20°C). Other solubilities: soluble in alcohol, insoluble in ether
Formula Weight 197.91
Percent Purity ≥99%
Grade Analytical
Packaging Plastic bottle
Chemical Name or Material Manganese(II) chloride tetrahydrate
Show More Show Less
Hazard Category GHS Signal Word: Danger
Hazard Statement GHS H Statement
May cause damage to organs through prolonged or repeated exposure if inhaled.
Harmful if swallowed.
Toxic to aquatic life with long lasting effects.
Causes serious eye damage.
Precautionary Statement GHS P Statement
IF SWALLOWED: rinse mouth.
Do NOT induce vomiting.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes.
Remove contact lenses,if pre

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.