Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Sigma Aldrich Manganese(II,III) oxide

Catalog No. 377473100G Shop All MilliporeSigma Sigma Organic Chemistry Products
Encompass
Encompass_Preferred
Change view
Click to view available options
Quantity:
100 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
37-747-3100G 100 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 37-747-3100G Supplier Sigma Aldrich Supplier No. 377473100G
Add to Cart
Add to Cart

MilliporeSigma Sigma Organics products encompass a wide range of quality reagents, solvents, catalysts, and building blocks for organic synthesis. From benchtop discovery to process development and scale-up, Sigma Organics solutions are built to meet the needs of synthetic chemists.

Manganese(II,III) oxide is a transition metal oxide that is formed by annealing manganese oxide in the air above 1000°C. It can be used for a variety of applications such as catalysis, electrochromic devices, and other energy storage applications.

Specifications

CAS 1317-35-7
Density 4.8 g/mL (at 25°C)
Molecular Formula Mn3O4
Linear Formula Mn3O4
Quantity 100 g
MDL Number MFCD00016218
Synonym MnO.Mn2O3
Molecular Weight (g/mol) 228.81
Percent Purity 97%
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.