Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Sigma Aldrich Manganese(II) nitrate hydrate

Catalog No. 2886401KG Shop All MilliporeSigma Sigma Organic Chemistry Products
Encompass
Encompass_Preferred
Change view
Click to view available options
Quantity:
25 g
500 g
1 kg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
28-864-01KG 1 kg
28-864-025G 25 g
28-864-0500G 500 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. 28-864-01KG Supplier Sigma Aldrich Supplier No. 2886401KG
May include imposed supplier surcharges.
Add to Cart
Add to Cart

MilliporeSigma Sigma Organics products encompass a wide range of quality reagents, solvents, catalysts, and building blocks for organic synthesis. From benchtop discovery to process development and scale-up, Sigma Organics solutions are built to meet the needs of synthetic chemists.

Manganese(II) nitrate hydrate is a low melting salt that can be used as a phase change material in thermal energy systems (TES).Manganese(II) nitrate hydrate can be used in the preparation of manganese doped cadmium selenide (CdSe) quantum dots. It can also be used in the synthesis of reactive ceramics for hydrogen generation based applications.

Specifications

CAS 15710-66-4
Quantity 1 kg
Molecular Formula MnN2O6 -+ xH2O
Linear Formula Mn(NO3)2-+xH2O
MDL Number MFCD00149788
Molecular Weight (g/mol) 178.95
Percent Purity 98%
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.