Learn More
Strem, An Ascensus Company CAS# 1344-43-0. 2kg. Manganese(II) oxide, 99%. MFCD00016226
SDP
Catalog No. 5090114599
Encompass
CAS# 1344-43-0. 2kg. Manganese(II) oxide, 99%. MFCD00016226. Molecular Weight: 70.94. Molecular Formula: MnO. Color/form: green pwdr. Strem# 93-2515. http://www.strem.com/catalog/v/93-2515/
Catalog No. 50-901-14599 Supplier Strem, An Ascensus Company Supplier No. 9325152000G
CAS# 1344-43-0. 2kg. Manganese(II) oxide, 99%. MFCD00016226. Molecular Weight: 70.94. Molecular Formula: MnO. Color/form: green pwdr. Strem# 93-2515. http://www.strem.com/catalog/v/93-2515/