Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Sigma Aldrich Manganese(II) phthalocyanine

Catalog No. 3795571G Shop All MilliporeSigma Sigma Organic Chemistry Products
Encompass
Change view
Click to view available options
Quantity:
1 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
37-955-71G 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 37-955-71G Supplier Sigma Aldrich Supplier No. 3795571G
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

MilliporeSigma Sigma Organics products encompass a wide range of quality reagents, solvents, catalysts, and building blocks for organic synthesis. From benchtop discovery to process development and scale-up, Sigma Organics solutions are built to meet the needs of synthetic chemists.

Manganese (II) phthalocyanine is used as a coating of quartz crystals which is to be used as sensors for detecting H2S. It is used to discriminate between cheese varieties, detect volatile hazardous compounds and spoilt fruit.

Specifications

CAS 14325-24-7
Molecular Formula C32H16MnN8
Linear Formula C32H16MnN8
MDL Number MFCD00049821
Quantity 1 g
Synonym Phthalocyanine manganese(II) salt
Molecular Weight (g/mol) 567.46
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.