Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Manganese(II) sulfate monohydrate, 102.0%, spray dried EMPROVE™ ESSENTIAL Ph Eur, USP, FCC, MilliporeSigma™

Catalog No. M1059991000 Shop All MilliporeSigma Products
Change view
Click to view available options
Quantity:
1 kg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
M1059991000 1 kg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. M1059991000 Supplier MilliporeSigma Supplier No. 1.05999.1000
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Spray dried, high-quality manganese(II) sulfate monohydrate designed for pharmaceutical solid formulation applications under the EMPROVE™ ESSENTIAL program.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 10034-96-5
Molecular Formula H2MnO5S
Molecular Weight (g/mol) 169.01
InChI Key ISPYRSDWRDQNSW-UHFFFAOYSA-L
IUPAC Name manganese(2+) hydrate sulfate
SMILES O.[Mn++].[O-]S([O-])(=O)=O

Specifications

Melting Point > 449°C (anhydrous substance)
pH 3.0 to 3.5 (50 g/L, H2O, 20°C)
Physical Form Fine Powder
Quantity 1 kg
Assay Percent Range Assay (complexometric): 98.0 to 102.0%; Assay (complexometric, calc. on the ignited basis): 99.0 to 101.0%
Solubility Information 762 g/L
Percent Purity 98.0 to 102.0% (complexometric)
Packaging Plastic Bottle
Density 2.95 g/cm3 (at 20°C); Bulk Density: 1000 to 1200 kg/m3
Chemical Name or Material Manganese(II) Sulfate
Recommended Storage Store at 15 to 25°C
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.