Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Manganese(II) sulfate tetrahydrate, 99% (metals basis)

Catalog No. aab2208122
Change view
Click to view available options
Quantity:
100 g
500 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAB2208122 100 g
AAB2208136 500 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAB2208122 Supplier Thermo Scientific Chemicals Supplier No. B2208122
Only null left
Add to Cart

CAS: 10101-68-5 | H8MnO8S | 223.054 g/mol

Manganese(II) sulfate tetrahydrate is an essential mineral used in capsule, tablet and liquid form, which provides essential nutrients like vitamin, protein and in similar nutritional substance. It undergoes electrolysis to give manganese dioxide. Upon oxidation, it gives chemical manganese dioxide (CMD) and finds application in dry- cell batteries.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Manganese(II) sulfate tetrahydrate is an essential mineral used in capsule, tablet and liquid form, which provides essential nutrients like vitamin, protein and in similar nutritional substance. It undergoes electrolysis to give manganese dioxide. Upon oxidation, it gives chemical manganese dioxide (CMD) and finds application in dry- cell batteries.

Solubility
Soluble in water.

Notes
Loses all water at 400-450°. Hygroscopic. Incompatible with strong oxidizing agents.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 10101-68-5
Molecular Formula H8MnO8S
Molecular Weight (g/mol) 223.054
MDL Number MFCD00149160
InChI Key CDUFCUKTJFSWPL-UHFFFAOYSA-L
Synonym manganese sulfate tetrahydrate, manganese ii sulfate tetrahydrate, unii-f46lh60l4m, manganese sulphate, tetrahydrate, mnso4.4h2o, acmc-1c6k2, mn.so4.4h2o, manganese 2+ sulfate tetrahydrate, manganese ii sulphate tetrahydrate
PubChem CID 22572548
ChEBI CHEBI:86358
IUPAC Name manganese(2+);sulfate;tetrahydrate
SMILES O.O.O.O.[O-]S(=O)(=O)[O-].[Mn+2]

Specifications

Quantity 100 g
Linear Formula MnSO4·4H2O
UN Number UN3077
Solubility Information Soluble in water.
Formula Weight 223.06 (151.00 Anhydrous)
Percent Purity 99%
Sensitivity Hygroscopic
Assay (metals basis)
Chemical Name or Material Manganese(II) sulfate tetrahydrate
Hazard Category Warning
Hazard Statement GHS H Statement
H373
May cause damage to organs through prolonged or repeated exposure.
Precautionary Statement GHS P Statement
P260-P314-P501a
Do not breathe dust/fume/gas/mist/vapors/spray.
Get medical advice/attention if you feel unwell.
Dispose of contents/container in accordance with local/regional/national/international regulations.
DOTInformation Transport Hazard Class: 9; Packing Group: III; Proper Shipping Name: ENVIRONMENTALLY HAZARDOUS SUBSTANCES, SOLID, N.O.S.
EINECSNumber 232-089-9
RTECSNumber OP1050000
TSCA Yes
Recommended Storage Ambient temperatures; Store under Nitrogen

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.