Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Manganese(III) 2,4-pentanedionate

Catalog No. p-7049024
Change view
Click to view available options
Quantity:
10 g
50 g
250 g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AA1253009 10 g
AA1253018 50 g
AA1253030 250 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. AA1253009 Supplier Thermo Scientific Chemicals Supplier No. 01253009
Only null left
Add to Cart

CAS: 14284-89-0 | C15H21MnO6 | 352.27 g/mol

It is mainly used as a solvent in paint and a precursor to vitamin E.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 14284-89-0
Molecular Formula C15H21MnO6
Molecular Weight (g/mol) 352.27
MDL Number MFCD00000023
InChI Key HYZQBNDRDQEWAN-LNTINUHCSA-K
Synonym mn iii acetylacetonate, tris acetylacetonato manganese iii
PubChem CID 122130696
IUPAC Name manganese(3+);(E)-4-oxopent-2-en-2-olate
SMILES [Mn+3].C\C([O-])=C\C(C)=O.C\C([O-])=C\C(C)=O.C\C([O-])=C\C(C)=O

Specifications

Melting Point ∼160°C (decomposition)
Quantity 10 g
Sensitivity Hygroscopic
Solubility Information Soluble in benzene, ethyl acetate
Formula Weight 352.24
Physical Form Powder
Chemical Name or Material Manganese(III) 2,4-pentanedionate
Hazard Category H302+H312+H332-H315-H319-H335
Precautionary Statement P261-P264b-P270-P271-P280-P301+P312-P302+P352-P304+P340-P305+P351+P338-P312-P330-P332+P313-P362-P501c
EINECSNumber 238-188-3
TSCA Yes
Recommended Storage Ambient temperatures

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.