Learn More
Strem, An Ascensus Company CAS# 19513-05-4. 5g. Manganese(III) acetate dihydrate, 98%. MFCD00150022
SDP
Catalog No. 5090110901
Encompass
CAS# 19513-05-4. 5g. Manganese(III) acetate dihydrate, 98%. MFCD00150022. Molecular Weight: 232.08 (268.10). Molecular Formula: Mn(OOCCH3)3∙2H2O. Color/form: brown xtl. Strem# 25-0750. http://www.strem.com/catalog/v/25-0750/
Catalog No. 50-901-10901 Supplier Strem, An Ascensus Company Supplier No. 2507505G
CAS# 19513-05-4. 5g. Manganese(III) acetate dihydrate, 98%. MFCD00150022. Molecular Weight: 232.08 (268.10). Molecular Formula: Mn(OOCCH3)3∙2H2O. Color/form: brown xtl. Strem# 25-0750. http://www.strem.com/catalog/v/25-0750/