Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Sigma Aldrich Manganese(III) Acetylacetonate

Catalog No. M2284100G Shop All MilliporeSigma Sigma Organic Chemistry Products
Encompass_Preferred
Change view
Click to view available options
Quantity:
10 mg
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
M2284100G 25 g
M228425G 10 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. M2284100G Supplier Sigma Aldrich Supplier No. M2284100G
  • NEW LOWER PRICE
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

MilliporeSigma Sigma Organics products encompass a wide range of quality reagents, solvents, catalysts, and building blocks for organic synthesis. From benchtop discovery to process development and scale-up, Sigma Organics solutions are built to meet the needs of synthetic chemists.

Manganese(III) Acetylacetonate

Specifications

CAS 14284-89-0
Melting Point 159°C to 161°C
Molecular Formula C15H21MnO6
Linear Formula Mn(C5H7O2)3
MDL Number MFCD00000023
Quantity 25 g
Synonym 2,4-Pentanedione manganese(III) derivative; Manganic acetylacetonate; Mn(acac)3
Molecular Weight (g/mol) 352.26 g/mol
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.