Learn More
Strem, An Ascensus Company CAS# 1313-13-9. 10g. Manganese(IV) oxide (99.995%-Mn) PURATREM. MFCD00003463
SDP
Catalog No. 5090110918
Encompass
CAS# 1313-13-9. 10g. Manganese(IV) oxide (99.995%-Mn) PURATREM. MFCD00003463. Molecular Weight: 86.94. Molecular Formula: MnO2. Color/form: black, random pieces Strem# 25-1380. http://www.strem.com/catalog/v/25-1380/
Catalog No. 50-901-10918 Supplier Strem, An Ascensus Company Supplier No. 25138010G
CAS# 1313-13-9. 10g. Manganese(IV) oxide (99.995%-Mn) PURATREM. MFCD00003463. Molecular Weight: 86.94. Molecular Formula: MnO2. Color/form: black, random pieces Strem# 25-1380. http://www.strem.com/catalog/v/25-1380/