Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Manganese(IV) oxide, 99.99+%, (trace metal basis), Thermo Scientific Chemicals

Catalog Number AC193470050
Click to view available options
Quantity:
5 g
25 g
100 g
Packaging:
Glass bottle
1313-13-9
MnO2
86.94
MFCD00003463
NUJOXMJBOLGQSY-UHFFFAOYSA-N
manganese dioxide, manganese iv oxide, manganese peroxide, manganese superoxide, manganese black, manganese binoxide, manganese oxide, mangandioxid, black manganese oxide, braunstein
14801
dioxomanganese
O=[Mn]=O
1313-13-9
MnO2
86.94
MFCD00003463
NUJOXMJBOLGQSY-UHFFFAOYSA-N
manganese dioxide, manganese iv oxide, manganese peroxide, manganese superoxide, manganese black, manganese binoxide, manganese oxide, mangandioxid, black manganese oxide, braunstein
14801
dioxomanganese
O=[Mn]=O

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

For ultra trace analysis, Oxidation reagent
TRUSTED_SUSTAINABILITY
Melting Point 535.0°C
Color Black to Gray
Physical Form Powder, Crystals and/or Chunks
Quantity 5 g
Assay Percent Range 99.99+%
Linear Formula MnO2
Fieser 01,637; 02,257; 03,191; 04,317; 05,422; 06,357; 08,312; 13,233; 14,200; 15,197; 16,202
Merck Index 15,5795
Solubility Information Solubility in water: insoluble. Other solubilities: soluble in hydrochloric acid
Formula Weight 86.94
Percent Purity ≥99.99%
Grade Trace Metal Basis
Packaging Glass bottle
Density 5.02g/mL
Chemical Name or Material Manganese(IV) oxide
Show More Show Less
Hazard Category GHS Signal Word: Warning
Hazard Statement GHS H Statement
Harmful if inhaled.
Harmful if swallowed.
May cause damage to organs through prolonged or repeated exposure if inhaled.
Precautionary Statement GHS P Statement
Wash face,hands and any exposed skin thoroughly after handling.
Call a POISON CENTER or doctor/physician if you feel unwell.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
D
EINECSNumber 215-202-6

RUO – Research Use Only

Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit