Learn More
Description
- Chemicals that do not have an established standard set for quality and impurity levels or for products where the purity is <90%
- Variation in color and physical form is possible
Specifications
Specifications
| Quantity | 250 g |
| Packaging | Plastic Bottle |
| UN Number | NONH for all modes of transport |
| Formula Weight | 86.94g/mol |
| Percent Purity | ≥90% |
| Grade | Technical |
| Chemical Name or Material | Manganese(IV) oxide activated |
Safety and Handling
| RTECSNumber | OP0350000 |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.