Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Manganese(IV) Oxide Powder, MilliporeSigma™

Catalog No. M1059571000 Shop All MilliporeSigma Products
Change view
Click to view available options
Quantity:
1 kg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
M1059571000 1 kg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. M1059571000 Supplier MilliporeSigma Supplier No. 1059571000
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

EMPLURA™ grade manganese(IV) oxide powder for laboratory applications.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 1313-13-9
Molecular Formula MnO2
Molecular Weight (g/mol) 86.94
MDL Number MFCD00003463
InChI Key NUJOXMJBOLGQSY-UHFFFAOYSA-N
Synonym manganese dioxide, manganese iv oxide, manganese peroxide, manganese superoxide, manganese black, manganese binoxide, manganese oxide, mangandioxid, black manganese oxide, braunstein
PubChem CID 14801
IUPAC Name dioxomanganese
SMILES O=[Mn]=O

Specifications

Physical Form Powder
Quantity 1 kg
UN Number UN 1479
Formula Weight 86.94
Percent Purity 89% (manganometric)
Odor Odorless
Packaging Glass bottle
Appearance Dark gray powder
Chemical Name or Material Manganese(IV) oxide
Hazard Category H272: May intensify fire; oxidiser.
H302 + H332: Harmful if swallowed or if inhaled
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.