Learn More
Hach Company Manganese LR PAN -50
Catalog No. 5000207037 Shop All Hach Company Products
Encompass
For determination of Manganese by the PAN Method using a 50-mm sample cell Hach Method 10286 Ultra Low Range 0 005 - 0 500 mg/L Mn 50 testsackage
Catalog No. 50-002-07037 Supplier Hach Company Supplier No. HPT291
For determination of Manganese by the PAN Method using a 50-mm sample cell Hach Method 10286 Ultra Low Range 0 005 - 0 500 mg/L Mn 50 testsackage