Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Sigma Aldrich Manganese Oxide (mno2) Nanosheets

Catalog No. 91804010ML Shop All MilliporeSigma Sigma Organic Chemistry Products
Encompass
Encompass_Preferred
Change view
Click to view available options
Quantity:
10 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
91-804-010ML 10 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 91-804-010ML Supplier Sigma Aldrich Supplier No. 91804010ML
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

MilliporeSigma Sigma Organics products encompass a wide range of quality reagents, solvents, catalysts, and building blocks for organic synthesis. From benchtop discovery to process development and scale-up, Sigma Organics solutions are built to meet the needs of synthetic chemists.

Transition metal oxides (TMOs, eg, MnO2), have received a substantial amount of recent attention due to their distinct structure-property relationships in multiple fields, eg, optoelectronics, spintronics, catalysis, defect engineering, and energy-related applications. Birnessite manganese dioxide (delta-MnO2) nanosheets are highly regarded as an attractive electrode material for portable energy storage devices. As the layered structure of delta-MnO2 enables much thinner and flexible devices.

Specifications

CAS 1313-13-9
Quantity 10 mL
Molecular Formula MnO2
Linear Formula MnO2
Synonym Birnessite manganese dioxide (delta-MnO2)
Molecular Weight (g/mol) 86.94
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.