Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Manganese Sulfate ACS AR Powder, Macron Fine Chemicals™

Catalog No. MK6192212 Shop All Avantor Products
Change view
Click to view available options
Poly; 4 x 2.5kg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No.
MK-6192-212 Poly; 4 x 2.5kg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. MK-6192-212 Supplier Avantor Supplier No. 619206

missing translation for 'validMainframeMsg'

Specifications

CAS 10034-96-5
Physical Form Powder
Quantity 4 x 2.5 kg
Molecular Formula H2MnO5S
MDL Number MFCD00149159
Synonym manganese sulfate monohydrate, manganese ii sulfate monohydrate, manganese sulfate hydrate, manganese ii sulfate hydrate, mnso4.h2o, manganous sulfate monohydrate, unii-w00lys4t26, manganese 2+ sulfate monohydrate, manganese sulfate usp, manganese 2+ sulfate hydrate
InChI Key ISPYRSDWRDQNSW-UHFFFAOYSA-L
SMILES O.[Mn++].[O-]S([O-])(=O)=O
IUPAC Name manganese(2+) hydrate sulfate
Molecular Weight (g/mol) 169.01
PubChem CID 177577
ChEBI CHEBI:86364
Formula Weight 169.02
Grade ACS
Packaging Poly
Chemical Name or Material Manganese Sulfate, Monohydrate
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.