Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Manganese Sulfate, Monohydrate, CP, Spectrum™ Chemical
SDP

Catalog No. 18611504 Shop All Spectrum Chemical Mfg. Corp. Products
Change view
Click to view available options
Quantity:
500 g
2.5 kg
12 kg
Packaging:
Poly Bottle
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
18-611-504 12 kg Poly Bottle
18-611-502 500 g Poly Bottle
18-611-503 2.5 kg Poly Bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. 18-611-504 Supplier Spectrum Chemical Manufacturing Corporation Supplier No. M111612KG
Get Up to $500 in Chemicals with Purchase
View all promotions
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

CAS: 10034-96-5 | 169.02 g/mol

Chemical Identifiers

CAS 10034-96-5

Specifications

CAS Max % 1
Physical Form Crystalline Powder or Needles
Quantity 12 kg
Molecular Weight (g/mol) 169.02
Grade Not Available
Packaging Poly Bottle
Chemical Name or Material Manganese Sulfate, Monohydrate

CAUTION: For manufacturing or laboratory use only. Read and understand the label and Safety Data Sheet (SDS) prior to use.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.