Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Manganese Sulfate, Monohydrate, Powder, USP, 98-102%, Spectrum™ Chemical
SDP

Catalog No. 18604208 Shop All Spectrum Chemical Mfg. Corp. Products
Change view
Click to view available options
Quantity:
500 g
2.5 kg
6 x 500 g
12 kg
4 x 2.5 kg
50 kg
Packaging:
Poly Bottle
6 product options available for selection
Product selection table with 6 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
18-604-208 500 g Poly Bottle
18-604-210 2.5 kg Poly Bottle
18-604-209 6 x 500 g Poly Bottle
18-604-211 4 x 2.5 kg Poly Bottle
18-604-212 12 kg Poly Bottle
18-604-213 50 kg Poly Bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 6 options available.
6 options
Catalog No. 18-604-208 Supplier Spectrum Chemical Manufacturing Corporation Supplier No. M1114500GM
Get Up to $500 in Chemicals with Purchase
View all promotions
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

M1114, 10034-96-5, MnSO4·H2O

Spectrum™ Chemical Manganese Sulfate, Monohydrate, Powder, USP - All Spectrum Chemical USP products are manufactured, packaged and stored under current Good Manufacturing Practices (cGMP) per 21CFR part 211 in FDA registered and inspected facilities.

Chemical Identifiers

CAS 10034-96-5
Molecular Formula H2MnO5S
Molecular Weight (g/mol) 169.01
MDL Number MFCD00149159
InChI Key ISPYRSDWRDQNSW-UHFFFAOYSA-L
IUPAC Name manganese(2+) hydrate sulfate
SMILES O.[Mn++].[O-]S([O-])(=O)=O

Specifications

Physical Form Fine Crystals or Crystalline Powder
CAS Max % 100%
Quantity 500 g
Loss on Ignition 10 to 13%
Packaging Poly Bottle
Percent Purity 98 to 102%
Grade USP
Chemical Name or Material Manganese Sulfate, Monohydrate, Powder
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.