Learn More
Chemscene AbaChemscene, Manganese triacetate dihydrate, 19513-05-4, 25g, Formula:C2H4O2.2/3H2O.1/3Mn, M. Wt. :268.1, Purity:>98%
SDP
Catalog No. 501635725
Encompass_Preferred
CS-W004700, AbaChemscene, Manganese triacetate dihydrate, 19513-05-4, 25g, Formula:C2H4O2.2/3H2O.1/3Mn, M. Wt. :268.1, Purity:>98%
Catalog No. 50-163-5725 Supplier Chemscene Supplier No. CSW00470025G
CS-W004700, AbaChemscene, Manganese triacetate dihydrate, 19513-05-4, 25g, Formula:C2H4O2.2/3H2O.1/3Mn, M. Wt. :268.1, Purity:>98%