Learn More
ALADDIN SCIENTIFIC CORPORATION ManganeseIII oxide, 100g
Supplier:  ALADDIN SCIENTIFIC CORPORATION M190551100G
Encompass_Preferred
Manganese(III) oxide (Mn2O3) can be used for a variety of potential applications such as supercapacitors fuel cells zinc ion batteries and lithium oxygen batteries Precursor to LiMn2O4 A useful battery cathode
Catalog No. 50-000-82503