Learn More
Sigma Aldrich Manganin™ - resistance alloy, Cu86Mn12Ni2
Catalog No. 503096875 Shop All MilliporeSigma Sigma Organic Chemistry Products
Click to view available options
Quantity:
1 Ea.
Catalog No. Quantity
50-309-6875 1 Ea.
1 options
Catalog No. 50-309-6875 Supplier Sigma Aldrich Supplier No. GF576475601EA
Add to Cart
Add to Cart
MilliporeSigma Sigma Organics products encompass a wide range of quality reagents, solvents, catalysts, and building blocks for organic synthesis. From benchtop discovery to process development and scale-up, Sigma Organics solutions are built to meet the needs of synthetic chemists.
Manganin™ - resistance alloy, Cu86Mn12Ni2
Specifications
Quantity 1 Ea.
Synonym CU066050