Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Fisher Science Education™ Manganous Chloride, Tetrahydrate
SCI_ED

Catalog No. S25418 Shop All Fisherbrand Products
Change view
Click to view available options
Quantity:
100 g
Packaging:
Reagent Grade
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
S25418 100 g Reagent Grade
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. S25418
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

This product is an Educational Grade Chemical — no C of A is available.

Chemical Identifiers

CAS 13446-34-9
Molecular Formula Cl2H8MnO4
Molecular Weight (g/mol) 197.90
MDL Number MFCD00149792
InChI Key CNFDGXZLMLFIJV-UHFFFAOYSA-L
Synonym Manganese (II) Chloride Tetrahydrate, Manganous Chloride Tetrahydrate
IUPAC Name manganese(2+) tetrahydrate dichloride
SMILES O.O.O.O.[Cl-].[Cl-].[Mn++]

Specifications

Quantity 100 g
Assay Percent Range >98%
Linear Formula MnCl2·4H2O
Formula Weight 197.91
Grade Reagent
Packaging Reagent Grade
Density 2.01g/mL
Chemical Name or Material Manganous Chloride, Tetrahydrate
Safety handling Check Alert storage code Red
ShelfLife 1 year
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.