Learn More
Hach Company Manganous Sulfate Powder Pillows, pk/50
Supplier:  Hach Company 107166
Encompass
For determination of Dissolved Oxygen by the Winkler titration method. Range: 1 to 10 mg/L. Hach methods 8215 and 8229. Sample size: 300 mL.
Catalog No. 50-199-4511