Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Manganous Sulfate Solution, LaMotte™
SDPSCI_ED

Catalog No. S72326 Shop All LaMotte Company Products
Change view
Click to view available options
Quantity:
30 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
S72326 30 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. S72326 Supplier LaMotte Company Supplier No. 4167G
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Reagent for dissolved oxygen water testing.

Chemical Identifiers

CAS 10034-96-5 , 7732-18-5
Synonym manganese sulfate monohydrate, manganese ii sulfate monohydrate, manganese sulfate hydrate, manganese ii sulfate hydrate, mnso4.h2o, manganous sulfate monohydrate, unii-w00lys4t26, manganese 2+ sulfate monohydrate, manganese sulfate usp, manganese 2+ sulfate hydrate

Specifications

Color Clear pink
Physical Form Liquid
Quantity 30 mL
Percent Purity 36%
Chemical Name or Material Manganous Sulfate Solution
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.