Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Manganous Sulfate Solution, 364 g/L, Ricca Chemical
SDP

Catalog No. R4620000100 Shop All Ricca Chemical Company Products
Change view
Click to view available options
Quantity:
100 mL
500 mL
1 L
Packaging:
Natural Poly Bottle
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
R4620000100 100 mL Natural Poly Bottle
4620-16 500 mL Natural Poly Bottle
4620-32 1 L Natural Poly Bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. R4620000100 Supplier Ricca Chemical Company Supplier No. 4620100
Add to Cart
Add to Cart

APHA Standard Methods Reagent

Specifications

Color Pink
CAS 7732-18-5, 100034-96-5, 7664-93-9
Boiling Point ∼100°C
CAS Min % 71.31, 28.66, 0.03
CAS Max % 71.31, 28.66, 0.03
Density 1.27
Synonym dihydrogen oxide, dihydrogen monoxide
Solubility Information Miscible in water
Molecular Weight (g/mol) 169.01 g/mol
Grade Laboratory
Packaging Natural Poly Bottle
Quantity 100 mL
Physical Form Liquid
Chemical Name or Material Manganous Sulfate Solution, 364 g/L
Show More Show Less
ShelfLife 24 months
Recommended Storage Ambient
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.