Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Manganous Sulfate Solution, 364 g/L, Ricca Chemical
SDP

Catalog No. 462032 Shop All Ricca Chemical Company Products
Change view
Click to view available options
Quantity:
100 mL
500 mL
1 L
Packaging:
Natural Poly Bottle
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
4620-32 1 L Natural Poly Bottle
R4620000100 100 mL Natural Poly Bottle
4620-16 500 mL Natural Poly Bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. 4620-32 Supplier Ricca Chemical Company Supplier No. 462032
Only null left
Add to Cart
Add to Cart

APHA Standard Methods Reagent

Chemical Identifiers

Molecular Formula H2MnO5S
MDL Number MFCD00149159
InChI Key ISPYRSDWRDQNSW-UHFFFAOYSA-L
PubChem CID 177577
ChEBI CHEBI:86364
IUPAC Name manganese(2+) hydrate sulfate
SMILES O.[Mn++].[O-]S([O-])(=O)=O

Specifications

Color Pink
CAS 10034-96-5, 7732-18-5
Synonym manganese sulfate monohydrate, manganese ii sulfate monohydrate, manganese sulfate hydrate, manganese ii sulfate hydrate, mnso4.h2o, manganous sulfate monohydrate, unii-w00lys4t26, manganese 2+ sulfate monohydrate, manganese sulfate usp, manganese 2+ sulfate hydrate
Molecular Weight (g/mol) 169.01
Grade Laboratory
Packaging Natural Poly Bottle
Quantity 1 L
Physical Form Liquid
Concentration or Composition Notes 364g/L
Chemical Name or Material Manganous Sulfate Solution
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.