Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Manganous Sulfate Solution APHA, ASTM for Dissolved Oxygen, Reagents

Catalog No. 01336472
Change view
Click to view available options
Quantity:
500 mL
1 L
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
01-336-472 1 L
01-336-471 500 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. 01-336-472 Supplier Reagents Holdings Llc Supplier No. C72430501A
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Manganous Sulfate Solution APHA, ASTM for Dissolved Oxygen, Reagent, for general laboratory use. Manufactured in ISO 9001 facility.

Specifications

Chemical Name or Material Manganous Sulfate Solution APHA, ASTM for Dissolved Oxygen
Boiling Point 103°C
Color Pink
CAS 7732-18-5, 10034-96-5
Physical Form Liquid
Quantity 1 L
Synonym Manganese Sulfate
Percent Purity 36.40%
Grade Reagent
Packaging Poly Bottle
Specific Gravity 1.27
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.