Learn More
Medchemexpress LLC HY-N0290 10mg Medchemexpress, Mangiferin CAS:4773-96-0 Purity:>98%
SDP
Catalog No. 501933509
Encompass_Preferred
Medchemexpress, HY-N0290 10mg Mangiferin CAS:4773-96-0 Mangiferin is a Nrf2 activator. Mangiferin suppresses nuclear translocation of the NF-κB subunits p65 and p50 . Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.
Catalog No. 50-193-3509 Supplier Medchemexpress LLC Supplier No. HYN029010MG
Medchemexpress, HY-N0290 10mg Mangiferin CAS:4773-96-0 Mangiferin is a Nrf2 activator. Mangiferin suppresses nuclear translocation of the NF-κB subunits p65 and p50 . Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.