Learn More
AdipoGen Mangiferin (1 g)
SDP
Supplier:  AdipoGen CDXM0465G001
Encompass_Preferred
Chemical. CAS: 4773-96-0. Formula: C19H18O11. MW: 422.34. Mangiferin is a xanthone glucoside that has been found in M. indica with diverse biological activities, including analgesic, antidiabetic, antisclerotic, antimicrobial and antiviral, cardio-, hepato-, and neuroprotective, anti-inflammatory, immunomodulatory, anticancer, as well as radioprotective against X-ray, gamma and UV radiation effects. It is also used in cosmetics, due to antioxidant and UV-protecting properties. The anti-inflammatroy activity is through inhibiting NF-kappa and NLRP3 signaling pathways.
Catalog No. 50-205-4541