Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Mangiferin, ≥98% (TLC), MilliporeSigma™ Supelco™

Catalog No. 111014250 Shop All MilliporeSigma Supelco Products
Change view
Click to view available options
Quantity:
10 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
11-101-4250 10 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 11-101-4250 Supplier MilliporeSigma Supelco Supplier No. 0627910MG
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Specifications

CAS 4773-96-0
Linear Formula C19H18O11
MDL Number MFCD00075656
Synonym 1,3,6,7-Tetrahydroxyxanthone C2-beta-D-glucoside; Chinomin; Chinonin; Shamimin
Type Analytical Standard
Quantity 10 mg
Formula Weight 422.34
Percent Purity ≥98% (TLC)
Grade Analytical Standard
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.