Learn More
Meridian Life Science MANGO MIX 250 REACTION
SDP
Catalog No. NC0059977
Encompass
MangoMix is a complete ready-to-go 2x reaction mix of the MangoTaq DNA Polymerase for carrying out primer extensions, requiring the user only to add water, template and primers.
Catalog No. NC0059977 Supplier Meridian Life Science Supplier No. BIO25033
MangoMix is a complete ready-to-go 2x reaction mix of the MangoTaq DNA Polymerase for carrying out primer extensions, requiring the user only to add water, template and primers.