Learn More
Thomas Scientific MangoMix, 2x (1000 x 50uL reactions)
SDP
Catalog No. NC1849429
Encompass
MangoMix is a convenient, ready-to-go, 2x Reaction Mix containing MangoTaq DNA Polymerase, MgCl2 and ultra-pure dNTPs manufactured by Bioline. The Mix is optimized and ready-to-use and requires only the addition of water, template and primers.
Catalog No. NC1849429 Supplier Thomas Scientific Supplier No. C755G90
MangoMix is a convenient, ready-to-go, 2x Reaction Mix containing MangoTaq DNA Polymerase, MgCl2 and ultra-pure dNTPs manufactured by Bioline. The Mix is optimized and ready-to-use and requires only the addition of water, template and primers.