Learn More
Cayman Chemical Mangostn 10mg
SDP
Catalog No. 502935184
Encompass_Preferred
A xanthone with diverse biological activities; inhibits growth of MRSA and MSSA (MICs = 6.25-12.5 and 6.25-25 μg/ml, respectively), P. falciparum (IC50 = 3 μg/ml), and M. tuberculosis (MIC = 6.25 μg/ml); inhibits FASN (IC50 = 24.83 μM); induces cell cycle arrest at the G2/M phase and intrinsic and mitochondrial apoptosis in MCF-7 breast cancer cells; inhibits LPS-induced nitric oxide and PGE2 production in RAW264.7 cells; reduces neutrophil infiltration and TNF-α and IL-1β production in a mouse model of carrageenan-induced peritonitis
Catalog No. 50-293-5184 Supplier Cayman Chemical Supplier No. 2507110 MG
A xanthone with diverse biological activities; inhibits growth of MRSA and MSSA (MICs = 6.25-12.5 and 6.25-25 μg/ml, respectively), P. falciparum (IC50 = 3 μg/ml), and M. tuberculosis (MIC = 6.25 μg/ml); inhibits FASN (IC50 = 24.83 μM); induces cell cycle arrest at the G2/M phase and intrinsic and mitochondrial apoptosis in MCF-7 breast cancer cells; inhibits LPS-induced nitric oxide and PGE2 production in RAW264.7 cells; reduces neutrophil infiltration and TNF-α and IL-1β production in a mouse model of carrageenan-induced peritonitis