Learn More
Apexbio Technology LLC Manidipine 89226-50-6 200mg
SDP
Catalog No. 5000062166
Encompass_Preferred
Manidipine (CAS 89226-50-6) is a dihydropyridine derivative functioning as a calcium channel blocker This compound specifically inhibits voltage-dependent calcium channels impairing calcium influx into vascular smooth muscle cells resulting in systemic arterial vasodilation Due to this mechanism manidipine is used mainly in research involving cardiovascular regulation hypertension models and investigations into calcium-mediated smooth muscle signaling pathways
Catalog No. 50-000-62166 Supplier Apexbio Technology LLC Supplier No. B1418200
Manidipine (CAS 89226-50-6) is a dihydropyridine derivative functioning as a calcium channel blocker This compound specifically inhibits voltage-dependent calcium channels impairing calcium influx into vascular smooth muscle cells resulting in systemic arterial vasodilation Due to this mechanism manidipine is used mainly in research involving cardiovascular regulation hypertension models and investigations into calcium-mediated smooth muscle signaling pathways