Learn More
Chem Service Inc Mandipropamid, 374726-62-2, MFCD09751510, 100 mg
SDP
Catalog No. NC1529046
Encompass_Preferred
Molecular Formula: C23H22ClNO4, Molecular Weight: 411.9, Classification: FUNGICIDE, Synonym: 2-(4-Chlorophenyl)-N-[3-methoxy-4-(2-propynyloxy)phenethyl]-2-(2-propynyloxy)acetamide. Mandipropamid is a carboxylic acid amide, which belongs to the oomycete class of fungicides. It is found to be effective in controlling phytopathogens such as Plasmopara viticola on grapes and potato late blight caused by Phytophthora infestans.
Catalog No. NC1529046 Supplier Chem Service Inc Supplier No. N12354100MG
Molecular Formula: C23H22ClNO4, Molecular Weight: 411.9, Classification: FUNGICIDE, Synonym: 2-(4-Chlorophenyl)-N-[3-methoxy-4-(2-propynyloxy)phenethyl]-2-(2-propynyloxy)acetamide. Mandipropamid is a carboxylic acid amide, which belongs to the oomycete class of fungicides. It is found to be effective in controlling phytopathogens such as Plasmopara viticola on grapes and potato late blight caused by Phytophthora infestans.